4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzyl alcohol

Lieferant: Apollo Scientific

Synonyme: [4-(3,5-Dimethyl-1H-pyrazol-1-yl)phenyl]methanol

OR14943-1G
APOSOR14943-1GEA 198 EUR
APOSOR14943-1G
4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzyl alcohol
4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzyl alcohol
Formel: C₁₂H₁₄N₂O
Molecular Weight: 202,252 g/mol
Schmelzpunkt: 57…58,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07324815
CAS-Nummer: 934570-55-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR