1-Methyl-1H-indole-7-carbonitrile 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Cyano-1-methyl-1H-indole

OR14945-100MG
APOSOR14945-100MGEA 87 EUR
APOSOR14945-100MG
1-Methyl-1H-indole-7-carbonitrile 97%
1-Methyl-1H-indole-7-carbonitrile
Formel: C₁₀H₈N₂
Molecular Weight: 156,184 g/mol
Schmelzpunkt: 67,5…69.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09263864
CAS-Nummer: 52951-14-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR