3-Amino-6-(phenylthio)pyridazine

Lieferant: Apollo Scientific
Achtung

OR14967-100MG
APOSOR14967-100MGEA 250 EUR
APOSOR14967-100MG
3-Amino-6-(phenylthio)pyridazine
3-Amino-6-(phenylthio)pyridazine
Formel: C₁₀H₉N₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD06801350
CAS-Nummer: 90844-35-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR