6-Phenyl-3(2H)-pyridazinon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Phenylpyridazin-3(2H)-one, 3-Hydroxy-6-phenylpyridazine
OR14974-5G
APOSOR14974-5GEA
55.1
EUR
APOSOR14974-5G
6-Phenyl-3(2H)-pyridazinon
6-Phenyl-3(2H)-pyridazinon
Formel:
C₁₀H₈N₂O Molecular Weight: 172,19 g/mol Schmelzpunkt: 201…204 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00114953 CAS-Nummer: 2166-31-6 EINECS: 218-505-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...