2-Amino-4-nitrobenzenemethanol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-4-nitrobenzyl alcohol
OR1499-250MG
APOSOR1499-250MGEA
21.9
EUR
APOSOR1499-250MG
2-Amino-4-nitrobenzenemethanol 98%
2-Amino-4-nitrobenzenemethanol
Formel:
C₇H₈N₂O₃ Molecular Weight: 168,1500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01664410 CAS-Nummer: 78468-34-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...