1,3-Oxazole-2-carbaldehyde 96%

Lieferant: Apollo Scientific
Achtung

OR14993-1G OR14993-250MG
APOSOR14993-1GEA 0 EUR
APOSOR14993-1G APOSOR14993-250MG
1,3-Oxazole-2-carbaldehyde 96%
1,3-Oxazole-2-carbaldehyde
Formel: C₄H₃NO₂
Lagertemperatur: Gefrierschrank
MDL: MFCD03412587
CAS-Nummer: 65373-52-6
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR