1,3-Oxazole-2-carbaldehyde 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Oxazole-2-carbaldehyde
OR14993-250MG
APOSOR14993-250MGEA
50.9
EUR
APOSOR14993-250MG
1,3-Oxazole-2-carbaldehyde 96%
1,3-Oxazole-2-carbaldehyde
Formel:
C₄H₃NO₂ Molecular Weight: 97,07 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD03412587 CAS-Nummer: 65373-52-6 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...