4-Brom-m-anisidin 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR15001-100G
OR15001-25G
OR15001-5G
APOSOR15001-100GEA
109
EUR
APOSOR15001-100G
APOSOR15001-25G
APOSOR15001-5G
4-Brom-m-anisidin 97%
4-Brom-m-anisidin
Formel:
C₇H₈BrNO Schmelzpunkt: 96…100 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD05664063 CAS-Nummer: 19056-40-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...