4-(tert-Butoxy)benzeneboronic acid 95+%

Lieferant: Apollo Scientific
Achtung

OR15015-1G
APOSOR15015-1GEA 62.5 EUR
APOSOR15015-1G
4-(tert-Butoxy)benzeneboronic acid 95+%
4-(tert-Butoxy)benzeneboronic acid
Formel: C₁₀H₁₅BO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03094965
CAS-Nummer: 176672-49-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR