Pyridiniumtribromid 90+%

Lieferant: Apollo Scientific
Gefahr

OR15023-100G OR15023-1KG OR15023-500G
APOSOR15023-100GEA 0 EUR
APOSOR15023-100G APOSOR15023-1KG APOSOR15023-500G
Pyridiniumtribromid 90+%
Pyridinium tribromide
Formel: C₅H₆Br₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD00013223
CAS-Nummer: 39416-48-3
EINECS: 254-446-8
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR