4-Biphenylboronsäurepinakolester 98%

Lieferant: Apollo Scientific
Achtung

OR15024-1G
APOSOR15024-1GEA 21.9 EUR
APOSOR15024-1G
4-Biphenylboronsäurepinakolester 98%
4-Biphenylboronsäurepinakolester
Formel: C₁₈H₂₁BO₂
Molecular Weight: 280.17 g/mol
Schmelzpunkt: 112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368291
CAS-Nummer: 144432-80-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR