N-[(3,5-Dimethylphenyl)carbamothioyl]benzamide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-[(Benzoylcarbamothioyl)amino]-m-xylene, 1-Benzoyl-3-(3,5-dimethylphenyl)thiourea
OR15025-5G
APOSOR15025-5GEA
169
EUR
APOSOR15025-5G
N-[(3,5-Dimethylphenyl)carbamothioyl]benzamide
N-[(3,5-Dimethylphenyl)carbamothioyl]benzamide
Formel:
C₁₆H₁₆N₂OS Molecular Weight: 284,376 g/mol Schmelzpunkt: 122…124 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02631870 CAS-Nummer: 117174-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...