4-Chlor-m-toluidin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Amino-2-chlortoluol, 4-Chlor-3-methylanilin
OR15033-100G
APOSOR15033-100GEA
109
EUR
APOSOR15033-100G
4-Chlor-m-toluidin 98%
4-Chlor-m-toluidin
Formel:
C₇H₈ClN Molecular Weight: 141,6 g/mol Schmelzpunkt: 82…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066332 CAS-Nummer: 7149-75-9 UN: 2239 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...