Ethyl 5-cyano-6-methyl-2-(piperazin-1-yl)nicotinate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 5-cyano-6-methyl-2-(piperazin-1-yl)pyridine-3-carboxylate, 3-Cyano-5-(ethoxycarbonyl)-2-methyl-6-(piperazin-1-yl)pyridine
OR15037-1G
APOSOR15037-1GEA
140
EUR
APOSOR15037-1G
Ethyl 5-cyano-6-methyl-2-(piperazin-1-yl)nicotinate
Ethyl 5-cyano-6-methyl-2-(piperazin-1-yl)nicotinate
Formel:
C₁₄H₁₈N₄O₂ Molecular Weight: 274.318 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01311995 CAS-Nummer: 402479-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...