2-Mercapto-1,3,4-thiadiazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3,4-Thiadiazole-2-thiol

OR15040-5G
APOSOR15040-5GEA 21.9 EUR
APOSOR15040-5G
2-Mercapto-1,3,4-thiadiazol
2-Mercapto-1,3,4-thiadiazole
Formel: C₂H₂N₂S₂
Molecular Weight: 118,17 g/mol
Schmelzpunkt: 142…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040404
CAS-Nummer: 18686-82-3
EINECS: 242-504-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR