1-(3,5-Dimethylphenyl)-2-thioharnstoff

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(3,5-Dimethylphenyl)thioharnstoff, 3,5-Dimethylphenylthioharnstoff, 1-(3,5-Xylyl)thioharnstoff

OR15045-500MG
APOSOR15045-500MGEA 21.9 EUR
APOSOR15045-500MG
1-(3,5-Dimethylphenyl)-2-thioharnstoff
1-(3,5-Dimethylphenyl)-2-thioharnstoff
Formel: (C₆H₃(CH₃)₂)NC(S)NH₂
Molecular Weight: 180,27 g/mol
Schmelzpunkt: 168…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041173
CAS-Nummer: 97480-60-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR