1-(4-Cyanphenyl)-2-thioharnstoff 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(4-Cyanophenyl)thiourea

OR15046-1G
APOSOR15046-1GEA 29 EUR
APOSOR15046-1G
1-(4-Cyanphenyl)-2-thioharnstoff 97%
1-(4-Cyanphenyl)-2-thioharnstoff
Formel: C₈H₇N₃S
Molecular Weight: 177,23 g/mol
Schmelzpunkt: 201…205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041175
CAS-Nummer: 3460-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR