Methyl-2-methyl-1,3-benzoxazole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

OR15047-250MG
APOSOR15047-250MGEA 29 EUR
APOSOR15047-250MG
Methyl-2-methyl-1,3-benzoxazole-5-carboxylate
Methyl-2-methyl-1,3-benzoxazole-5-carboxylate
Formel: C₁₀H₉NO₃
Molecular Weight: 191.19 g/mol
Schmelzpunkt: 66…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07774245
CAS-Nummer: 136663-21-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR