Methyl-2-propyl-1,3-benzoxazole-5-carboxylate

Lieferant: Apollo Scientific

OR15050-1G
APOSOR15050-1GEA 0 EUR
APOSOR15050-1G
Methyl-2-propyl-1,3-benzoxazole-5-carboxylate
Methyl-2-propyl-1,3-benzoxazole-5-carboxylate
Formel: C₁₂H₁₃NO₃
Molecular Weight: 219,24 g/mol
Schmelzpunkt: 48…50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689718
CAS-Nummer: 924869-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR