4-Amino-3-penten-2-on 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Amino-3-penten-2-one, 4-Aminopent-3-en-2-one
OR1506-100G
APOSOR1506-100GEA
177
EUR
APOSOR1506-100G
4-Amino-3-penten-2-on 95%
Fluoral-P (=4-amino-3-penten-2-one)
Formel:
C₅H₉NO Siedepunkt: 130…131 °C Schmelzpunkt: 38 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00043715 CAS-Nummer: 1118-66-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...