3-[4-(1,3-Benzoxazol-2-yl)piperidino]propanoic acid

Lieferant: Apollo Scientific
Achtung

OR15064-100MG
APOSOR15064-100MGEA 26.1 EUR
APOSOR15064-100MG
3-[4-(1,3-Benzoxazol-2-yl)piperidino]propanoic acid
3-[4-(1,3-Benzoxazol-2-yl)piperidino]propanoic acid
Formel: C₁₅H₁₈N₂O₃
Molecular Weight: 274,32 g/mol
Schmelzpunkt: 185…187 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08444012
CAS-Nummer: 924869-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR