4-Chloro-3-methyl-5-isoxazolamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-4-chloro-3-methylisoxazole

OR15076-250MG
APOSOR15076-250MGEA 21.9 EUR
APOSOR15076-250MG
4-Chloro-3-methyl-5-isoxazolamine
4-Chloro-3-methyl-5-isoxazolamine
Formel: C₄H₅ClN₂O
Molecular Weight: 132,55 g/mol
Schmelzpunkt: 65…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08443950
CAS-Nummer: 166964-09-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR