4-Phthalimidobutyric acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(1,3-Dioxoisoindolin-2-yl)butanoic acid, 4-(Phthalimid-1-yl)butanoic acid
OR15083-1G
OR15083-250MG
APOSOR15083-1GEA
63.8
EUR
APOSOR15083-1G
APOSOR15083-250MG
4-Phthalimidobutyric acid
4-Phthalimidobutyric acid
Formel:
C₁₂H₁₁NO₄ Schmelzpunkt: 119…121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00196079 CAS-Nummer: 3130-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...