Ethyl-6-brom-4-hydroxychinolin-3-carboxylat 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 6-bromo-4-hydroxyquinoline-3-carboxylate
OR15084-25G
APOSOR15084-25GEA
171
EUR
APOSOR15084-25G
Ethyl-6-brom-4-hydroxychinolin-3-carboxylat 95%
Ethyl-6-brom-4-hydroxychinolin-3-carboxylat
Formel:
C₁₂H₁₀BrNO₃ Molecular Weight: 296.1200 g/mol Schmelzpunkt: 326…329 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173362 CAS-Nummer: 122794-99-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...