Ethyl 2-cyano-3-(4-hydroxyphenyl)acrylate

Lieferant: Apollo Scientific
Achtung

Synonyme: (E)-Ethyl 2-cyano-3-(4-hydroxyphenyl)acrylate

OR15088-1G
APOSOR15088-1GEA 34.8 EUR
APOSOR15088-1G
Ethyl 2-cyano-3-(4-hydroxyphenyl)acrylate
Ethyl-2-cyano-3-(4-hydroxyphenyl)acrylate
Formel: C₁₂H₁₁NO₃
Schmelzpunkt: 180…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00188426
CAS-Nummer: 6935-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR