Ethyl-4-hydroxy-2-(4-methoxyphenyl)-1,3-thiazole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

OR15092-500MG
APOSOR15092-500MGEA 48 EUR
APOSOR15092-500MG
Ethyl-4-hydroxy-2-(4-methoxyphenyl)-1,3-thiazole-5-carboxylate
Ethyl-4-hydroxy-2-(4-methoxyphenyl)-1,3-thiazole-5-carboxylate
Formel: C₁₃H₁₃NO₄S
Molecular Weight: 279,32 g/mol
Schmelzpunkt: 147…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08444006
CAS-Nummer: 924869-01-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR