Ethyl-4-hydroxy-2-(2-thienyl)-1,3-thiazole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

OR15093-250MG
APOSOR15093-250MGEA 30.6 EUR
APOSOR15093-250MG
Ethyl-4-hydroxy-2-(2-thienyl)-1,3-thiazole-5-carboxylate
Ethyl-4-hydroxy-2-(2-thienyl)-1,3-thiazole-5-carboxylate
Formel: C₁₀H₉NO₃S₂
Molecular Weight: 255,32 g/mol
Schmelzpunkt: 128…130 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08444005
CAS-Nummer: 924869-00-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR