Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 4-Quinazolone-2-carboxylate, 4-Quinazolone-2-carboxylic acid ethyl ester

OR15094-1G
APOSOR15094-1GEA 0 EUR
APOSOR15094-1G
Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate
Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate
Formel: C₁₁H₁₀N₂O₃
Molecular Weight: 218,21 g/mol
Schmelzpunkt: 189…193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051583
CAS-Nummer: 29113-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR