Methyl-3-chloro-1H-indole-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR15103-100MG
APOSOR15103-100MGEA 43.5 EUR
APOSOR15103-100MG
Methyl-3-chloro-1H-indole-2-carboxylate
Methyl-3-chloro-1H-indole-2-carboxylate
Formel: C₁₀H₈ClNO₂
Molecular Weight: 209,63 g/mol
Schmelzpunkt: 168…170 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04125889
CAS-Nummer: 220664-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR