3-Oxo-3-(2-pyridinyl)propanenitrile 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Oxo-3-(pyridin-2-yl)propanenitrile

OR15118-100MG
APOSOR15118-100MGEA 21.9 EUR
APOSOR15118-100MG
3-Oxo-3-(2-pyridinyl)propanenitrile 95%
3-Oxo-3-(2-pyridinyl)propanenitrile
Formel: C₈H₆N₂O
Molecular Weight: 146,15 g/mol
Schmelzpunkt: 97…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06656806
CAS-Nummer: 54123-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR