2-Phenyl-1,3-thiazol-4-ol
Lieferant: Apollo Scientific
|
Achtung
|
OR15120-50MG
APOSOR15120-50MGEA
29
EUR
APOSOR15120-50MG
2-Phenyl-1,3-thiazol-4-ol
2-Phenyl-1,3-thiazol-4-ol
Formel:
C₉H₇NOS Schmelzpunkt: 119…121 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08444022 CAS-Nummer: 827-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...