4-(4'-Piperid-1-yl)-1-tert-butoxycarbonylpiperidine

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl [4,4'-bipiperidine]-1-carboxylate, 4,4'-Bipiperidine, 1-BOC protected, N-Boc-4,4'-bipiperidine

OR15121-1G
APOSOR15121-1GEA 0 EUR
APOSOR15121-1G
4-(4'-Piperid-1-yl)-1-tert-butoxycarbonylpiperidine
4-(4'-Piperid-1-yl)-1-tert-butoxycarbonylpiperidine
Formel: C₁₅H₂₈N₂O₂
Molecular Weight: 268,4 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02179169
CAS-Nummer: 171049-35-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR