Potassium-4-iodobenzenesulphonate

Lieferant: Apollo Scientific
Achtung

Synonyme: Potassium 4-iodobenzenesulphonate

OR15122-1G OR15122-5G
APOSOR15122-1GEA 126 EUR
APOSOR15122-1G APOSOR15122-5G
Potassium-4-iodobenzenesulphonate
Potassium-4-iodobenzenesulphonate
Formel: C₆H₄IKO₃S
Molecular Weight: 322.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06656232
CAS-Nummer: 13035-63-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR