tert-Butyl-4-[3-(1H-pyrazol-5-yl)phenethyl]tetrahydro-1(2H)-pyrazinecarboxylate

Lieferant: Apollo Scientific

Synonyme: 4-[3-(1H-Pyrazol-5-yl)phenethyl]piperazine, N1-BOC protected

OR15123-500MG
APOSOR15123-500MGEA 438 EUR
APOSOR15123-500MG
tert-Butyl-4-[3-(1H-pyrazol-5-yl)phenethyl]tetrahydro-1(2H)-pyrazinecarboxylate
tert-Butyl-4-[3-(1H-pyrazol-5-yl)phenethyl]tetrahydro-1(2H)-pyrazinecarboxylate
Formel: C₂₀H₂₈N₄O₂
Molecular Weight: 356,47 g/mol
Schmelzpunkt: 88…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08444013
CAS-Nummer: 1186648-43-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR