2-Amino-5-phenyl-1,3-thiazol-4-ol

Lieferant: Apollo Scientific
Achtung

OR15127-100MG
APOSOR15127-100MGEA 21.9 EUR
APOSOR15127-100MG
2-Amino-5-phenyl-1,3-thiazol-4-ol
2-Amino-5-phenyl-1,3-thiazol-4-ol
Formel: C₉H₈N₂OS
Molecular Weight: 192,24 g/mol
Schmelzpunkt: 240…242 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08444003
CAS-Nummer: 98879-58-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR