4-Bromo-2-chloro-6-[(dimethylamino)methyl]benzenol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-chloro-6-[(dimethylamino)methyl]phenol

OR15134-1G
APOSOR15134-1GEA 180 EUR
APOSOR15134-1G
4-Bromo-2-chloro-6-[(dimethylamino)methyl]benzenol
4-Bromo-2-chloro-6-[(dimethylamino)methyl]benzenol
Formel: C₉H₁₁BrClNO
Molecular Weight: 264,56 g/mol
Schmelzpunkt: 69…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08443982
CAS-Nummer: 924868-93-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR