2-Brom-1-(2-thienyl)-1-propanon

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-1-(thiophen-2-yl)propan-1-one

OR15139-250MG
APOSOR15139-250MGEA 21.9 EUR
APOSOR15139-250MG
2-Brom-1-(2-thienyl)-1-propanon
2-Brom-1-(2-thienyl)-1-propanon
Formel: C₇H₇BrOS
Molecular Weight: 219,0989 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08443970
CAS-Nummer: 75815-46-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR