2-Chlor-6-iodanilin 98+%

Lieferant: Apollo Scientific
Achtung

OR15148-1G
APOSOR15148-1GEA 36.4 EUR
APOSOR15148-1G
2-Chlor-6-iodanilin 98+%
2-Chlor-6-iodanilin
Formel: C₆H₅ClIN
Molecular Weight: 253.4681 g/mol
Schmelzpunkt: 64…67 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08443948
CAS-Nummer: 84483-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR