4-(4-Chlorobenzoyl)-1H-pyrrole-2-carboxylic acid

Lieferant: Apollo Scientific

OR15150-1G
APOSOR15150-1GEA 336 EUR
APOSOR15150-1G
4-(4-Chlorobenzoyl)-1H-pyrrole-2-carboxylic acid
4-(4-Chlorobenzoyl)-1H-pyrrole-2-carboxylic acid
Formel: C₁₂H₈ClNO₃
Schmelzpunkt: 248…251 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08443966
CAS-Nummer: 72531-60-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR