4-Amino-3-methylbenzolsulfonamid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR1516-1G
APOSOR1516-1GEA
62.5
EUR
APOSOR1516-1G
4-Amino-3-methylbenzolsulfonamid 95%
4-Amino-3-methylbenzolsulfonamid
Formel:
C₇H₁₀N₂O₂S Molecular Weight: 186,2300 g/mol Schmelzpunkt: 160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00466840 CAS-Nummer: 53297-70-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...