3-(3,4-Dimethoxyphenyl)-1H-pyrazol-5-amin

Lieferant: Apollo Scientific
Achtung

OR15171-1G
APOSOR15171-1GEA 108 EUR
APOSOR15171-1G
3-(3,4-Dimethoxyphenyl)-1H-pyrazol-5-amin
3-(3,4-Dimethoxyphenyl)-1H-pyrazol-5-amin
Formel: C₁₁H₁₃N₃O₂
Schmelzpunkt: 131…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02664228
CAS-Nummer: 208519-08-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR