Ethyl-2-chloro-6-benzothiazolecarboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2-chloro-6-benzothiazolecarboxylate
OR15184-5G
APOSOR15184-5GEA
98.6
EUR
APOSOR15184-5G
Ethyl-2-chloro-6-benzothiazolecarboxylate
Ethyl-2-chloro-6-benzothiazolecarboxylate
Formel:
C₁₀H₈ClNO₂S Molecular Weight: 241.6900 g/mol Schmelzpunkt: 90…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08443995 CAS-Nummer: 78485-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...