Ethyl-4-(4,4-dimethyl-2,6-dioxopiperidino)benzenecarboxylate

Lieferant: Apollo Scientific

OR15185-10G OR15185-1G OR15185-5G
APOSOR15185-10GEA 383 EUR
APOSOR15185-10G APOSOR15185-1G APOSOR15185-5G
Ethyl-4-(4,4-dimethyl-2,6-dioxopiperidino)benzenecarboxylate
Ethyl-4-(4,4-dimethyl-2,6-dioxopiperidino)benzenecarboxylate
Formel: C₁₆H₁₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD01764136
CAS-Nummer: 279692-23-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR