6-Ethyl-5,7-dimethyl[1,2,4]triazolo[1,5-a]pyrimidin-2-amine
Lieferant: Apollo Scientific
Synonyme:
2-Amino-6-ethyl-5,7-dimethyl[1,2,4]triazolo[1,5-a]pyrimidine
OR15186-1G
APOSOR15186-1GEA
270
EUR
APOSOR15186-1G
6-Ethyl-5,7-dimethyl[1,2,4]triazolo[1,5-a]pyrimidin-2-amine
6-Ethyl-5,7-dimethyl[1,2,4]triazolo[1,5-a]pyrimidin-2-amine
Formel:
C₉H₁₃N₅ Molecular Weight: 191.24 g/mol Schmelzpunkt: 297…300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07402021 CAS-Nummer: 924868-96-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...