Ethyl-4-(2-hydroxy-5-methyl-3-nitrophenyl)-2,4-dioxobutanoate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2,4-dioxo-4-(2-hydroxy-5-methyl-3-nitrophenyl)butanoate
OR15192-1G
APOSOR15192-1GEA
50.9
EUR
APOSOR15192-1G
Ethyl-4-(2-hydroxy-5-methyl-3-nitrophenyl)-2,4-dioxobutanoate
Ethyl-4-(2-hydroxy-5-methyl-3-nitrophenyl)-2,4-dioxobutanoate
Formel:
C₁₃H₁₃NO₇ Molecular Weight: 295,25 g/mol Schmelzpunkt: 116…118 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08443987 CAS-Nummer: 859843-60-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...