Ethyl-2-(1H-pyrazol-1-yl)acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-(1-Pyrazolyl)acetate

OR15197-250MG
APOSOR15197-250MGEA 36.4 EUR
APOSOR15197-250MG
Ethyl-2-(1H-pyrazol-1-yl)acetate
Ethyl-2-(1H-pyrazol-1-yl)acetate
Formel: C₇H₁₀N₂O₂
Molecular Weight: 154,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00159620
CAS-Nummer: 10199-61-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR