5-Amino-3-(3-methoxyphenyl)isoxazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(5-Amino-1,2-oxazol-3-yl)anisole, 3-(3-Methoxyphenyl)isoxazol-5-amine

OR15205-1G
APOSOR15205-1GEA 21.9 EUR
APOSOR15205-1G
5-Amino-3-(3-methoxyphenyl)isoxazole
5-Amino-3-(3-methoxyphenyl)isoxazole
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,199 g/mol
Schmelzpunkt: 91…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07383035
CAS-Nummer: 119162-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR