4-[(3-Formyl-2-pyridinyl)oxy]benzenecarbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-[(3-Formylpyridin-2-yl)oxy]benzonitrile
OR15206-100MG
APOSOR15206-100MGEA
21.9
EUR
APOSOR15206-100MG
4-[(3-Formyl-2-pyridinyl)oxy]benzenecarbonitrile
4-[(3-Formyl-2-pyridinyl)oxy]benzenecarbonitrile
Formel:
C₁₃H₈N₂O₂ Molecular Weight: 224.22 g/mol Schmelzpunkt: 162…166 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03305730 CAS-Nummer: 866049-85-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...