3,4-Dihydro-5-hydroxy-1H-quinolin-2-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Hydroxy-2-oxo-1,2,3,4-tetrahydroquinoline, 5-Hydroxy-3,4-dihydro-2(1H)-quinolinone
OR15207-250MG
APOSOR15207-250MGEA
60.9
EUR
APOSOR15207-250MG
3,4-Dihydro-5-hydroxy-1H-quinolin-2-one
3,4-Dihydro-5-hydroxy-1H-quinolin-2-one
Formel:
C₉H₉NO₂ Molecular Weight: 163,18 g/mol Schmelzpunkt: 225…227 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862194 CAS-Nummer: 30389-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...