2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR15224-1G
APOSOR15224-1GEA 92.8 EUR
APOSOR15224-1G
2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid
2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid
Formel: C₁₀H₉NO₄
Molecular Weight: 207,19 g/mol
Schmelzpunkt: 192…194 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04124544
CAS-Nummer: 154365-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR